Gene Transcripts

Gene Symbol
POMGNT2
Entrez Gene ID
84892
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 2 (beta 1,4-)
Gene Aliases
AGO61, C3orf39, GTDC2, MDDGA8
Chromosome Location
Chr. 3: 43079229 - 43106092 on Build GRCh38
UniGene ID
Hs.745002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032806.5 NP_116195.2 2 859
XM_005265515.2 XP_005265572.1 3 990
XM_011534163.2 XP_011532465.1 3 978
XM_011534164.1 XP_011532466.1 2 1560
XM_017007353.1 XP_016862842.1 4 1069
GenBank mRNA AK027472.1 - 1 119
AK092147.1 - 2 647
AK124737.1 - 1 1827
BC060861.1 - 2 849 30335722

View Full Product Details