Gene Transcripts

Gene Symbol
AGK
Entrez Gene ID
55750
Gene Name
acylglycerol kinase
Gene Aliases
CATC5, CTRCT38, MTDPS10, MULK
Chromosome Location
Chr. 7: 141551278 - 141655244 on Build GRCh38
UniGene ID
Hs.743318
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018238.3 NP_060708.1 16 2896
XM_011516397.2 XP_011514699.1 16 3362
GenBank mRNA BC022777.1 - 15 2750

View Full Product Details