Gene Transcripts

Gene Symbol
MAGIX
Entrez Gene ID
79917
Gene Name
MAGI family member, X-linked
Gene Aliases
JM10, PDZX
Chromosome Location
Chr. X: 49162569 - 49168157 on Build GRCh38
UniGene ID
Hs.193170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024859.3 NP_079135.3 3, 4 334
XM_005278064.4 XP_005278121.1 2 267
XM_005278065.3 XP_005278122.1 2 355
XM_006724560.3 XP_006724623.1 3, 4 418
XM_011543985.2 XP_011542287.1 3 488
XM_011543986.2 XP_011542288.1 2 242
XM_011543987.2 XP_011542289.1 2 263
GenBank mRNA AK025340.1 - 2 189
AK096058.1 - 2 1429
DA072017.1 - 2, 3 116
DQ884401.1 - 1 110

View Full Product Details