Gene Transcripts

Gene Symbol
C5orf24
Entrez Gene ID
134553
Gene Name
chromosome 5 open reading frame 24
Gene Aliases
Chromosome Location
Chr. 5: 134833603 - 134859735 on Build GRCh38
UniGene ID
Hs.708536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135586.1 NP_001129058.1 2 787
NM_152409.3 NP_689622.2 2 600
XM_005271889.3 XP_005271946.1 2 1436
XM_017009049.1 XP_016864538.1 2 955
XM_017009050.1 XP_016864539.1 2 563
GenBank mRNA AK094881.1 - 2 737
BC053677.1 - 2 600 6166677
BX648529.1 - 2 667

View Full Product Details