Gene Transcripts

Gene Symbol
ATXN7L1
Entrez Gene ID
222255
Gene Name
ataxin 7 like 1
Gene Aliases
ATXN7L4
Chromosome Location
Chr. 7: 105604774 - 105877232 on Build GRCh38
UniGene ID
Hs.745056
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152749.3 NP_689962.1 4 632
XM_011515954.2 XP_011514256.1 5 715
GenBank mRNA AK095475.1 - 4 546
BC030616.2 - 4 629 4796569

View Full Product Details