Gene Transcripts

Gene Symbol
BEND2
Entrez Gene ID
139105
Gene Name
BEN domain containing 2
Gene Aliases
CXorf20
Chromosome Location
Chr. X: 18162931 - 18220904 on Build GRCh38
UniGene ID
Hs.403802
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153346.4 NP_699177.2 14 3178
GenBank mRNA AK128155.1 - 14 3159
BC037301.2 - 14 3157 4827886

View Full Product Details