Gene Transcripts

Gene Symbol
ERICH5
Entrez Gene ID
203111
Gene Name
glutamate rich 5
Gene Aliases
C8orf47
Chromosome Location
Chr. 8: 98064522 - 98093610 on Build GRCh38
UniGene ID
Hs.171455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173549.2 NP_775820.2 2 573
GenBank mRNA AK096872.1 - 2 573
BC062359.1 - 2 517 5015294

View Full Product Details