Gene Transcripts

Gene Symbol
AMOT
Entrez Gene ID
154796
Gene Name
angiomotin
Gene Aliases
Chromosome Location
Chr. X: 112774877 - 112840946 on Build GRCh38
UniGene ID
Hs.528051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113490.1 NP_001106962.1 11 3739
NM_133265.2 NP_573572.1 12 3308
XM_005262087.1 XP_005262144.1 14 4484
XM_005262090.1 XP_005262147.1 11 2981
XM_011530875.2 XP_011529177.1 15 4596
XM_017029289.1 XP_016884778.1 13 3512
GenBank mRNA AB028994.3 - 10 2512
AF286598.1 - 12 3308
BC130294.1 - 10 2732 40146325

View Full Product Details