Gene Transcripts

Gene Symbol
NUS1
Entrez Gene ID
116150
Gene Name
NUS1 dehydrodolichyl diphosphate synthase subunit
Gene Aliases
C6orf68, MGC:7199, NgBR, TANGO14
Chromosome Location
Chr. 6: 117675454 - 117710727 on Build GRCh38
UniGene ID
Hs.289008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138459.3 NP_612468.1 5 1658
GenBank mRNA AK125119.1 - 5 1620
AK126335.1 - 6 1372
AK130093.1 - 4 975
BC013026.2 - 5 1605 3531630
BC063794.1 - 5 1612 6500832
BC066910.1 - 5 1652 4830860
BC110325.1 - 5 1614 5553827

Gene Symbol
LOC105370109
Entrez Gene ID
105370109
Gene Name
uncharacterized LOC105370109
Gene Aliases
Chromosome Location
Chr. 13: 22883682 - 22909059 on Build GRCh38
UniGene ID
Hs.380803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK130366.1 - 4 640

View Full Product Details