Gene Transcripts
- Gene Symbol
- SPATS2
- Entrez Gene ID
- 65244
- Gene Name
- spermatogenesis associated serine rich 2
- Gene Aliases
- Nbla00526, P59SCR, SCR59, SPATA10
- Chromosome Location
- Chr. 12: 49366905 - 49527441 on Build GRCh38
- UniGene ID
- Hs.654826
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293285.1 |
NP_001280214.1
|
10, 11 |
1917 |
|
NM_001293286.1 |
NP_001280215.1
|
8, 9 |
1770 |
|
NM_023071.3 |
NP_075559.2
|
9, 10 |
1833 |
|
XM_011538690.2 |
XP_011536992.1
|
10, 11 |
1305 |
|
XM_011538691.2 |
XP_011536993.1
|
8, 9 |
1200 |
|
XM_017019840.1 |
XP_016875329.1
|
9, 10 |
1273 |
|
GenBank mRNA |
AB075515.1 |
- |
7, 8 |
861 |
|
AK023179.1 |
- |
5, 6 |
742 |
|
AK023202.1 |
- |
9, 10 |
1232 |
|
AK024678.1 |
- |
1, 2 |
129 |
|
AK292334.1 |
- |
8, 9 |
1170 |
|
AL833614.1 |
- |
10, 11 |
1358 |
|
BC048299.1 |
- |
8, 9 |
1200 |
5520818 |
BX648916.1 |
- |
8, 9 |
1143 |
|
View Full Product Details