Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
79786
Gene Name
kelch like family member 36
Gene Aliases
C16orf44
Chromosome Location
Chr. 16: 84648504 - 84667686 on Build GRCh38
UniGene ID
Hs.222731
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303451.1 NP_001290380.1 4 1360
NM_024731.3 NP_079007.2 5 1549
XM_005256149.2 XP_005256206.1 5 1763
XM_017023674.1 XP_016879163.1 5 1511
XM_017023675.1 XP_016879164.1 5 1531
GenBank mRNA AK022605.1 - 4 1402
AK074056.1 - 2 478
AK122745.1 - 3 1984
BC032400.1 - 4 1336 5210986
BC033821.1 - 5 1535 5219021

View Full Product Details