Gene Transcripts

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
Chromosome Location
Chr. 6: 108848410 - 108990738 on Build GRCh38
UniGene ID
Hs.645481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286609.1 NP_001273538.1 13 2072
NM_032131.5 NP_115507.4 14 2145
XM_005267154.3 XP_005267211.1 14 2426
XM_005267155.2 XP_005267212.1 14 2161
XM_005267157.3 XP_005267214.1 14 2069
XM_006715573.2 XP_006715636.1 15 2251
XM_006715574.2 XP_006715637.1 11 1581
XM_011536166.1 XP_011534468.1 15 2236
XM_011536167.1 XP_011534469.1 14 2022
XM_011536168.2 XP_011534470.1 14 2139
XM_011536170.2 XP_011534472.1 14 2139
XM_011536172.2 XP_011534474.1 13 1996
XM_017011350.1 XP_016866839.1 13 2074
XM_017011352.1 XP_016866841.1 13 1975
GenBank mRNA AK095234.1 - 13 1988
AK292499.1 - 14 2139
AK294574.1 - 13 2072
AL136754.1 - 10 1495
BC030603.2 - 14 2139 4824783

View Full Product Details