Gene Transcripts

Gene Symbol
CARD6
Entrez Gene ID
84674
Gene Name
caspase recruitment domain family member 6
Gene Aliases
CINCIN1
Chromosome Location
Chr. 5: 40841190 - 40855354 on Build GRCh38
UniGene ID
Hs.200242
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032587.3 NP_115976.2 2 689
XM_017009990.1 XP_016865479.1 2 807
GenBank mRNA AB209844.1 - 2 509
AF356193.1 - 2 813
AK304349.1 - 2 621
AY196783.1 - 2 689
BC093825.1 - 2 636 7939670

View Full Product Details