Gene Transcripts

Gene Symbol
ADAM12
Entrez Gene ID
8038
Gene Name
ADAM metallopeptidase domain 12
Gene Aliases
ADAM12-OT1, CAR10, MCMP, MCMPMltna, MLTN, MLTNA
Chromosome Location
Chr. 10: 126012385 - 126388558 on Build GRCh38
UniGene ID
Hs.594351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288973.1 NP_001275902.1 2 530
NM_001288974.1 NP_001275903.1 2 530
NM_001288975.1 NP_001275904.1 2 530
NM_003474.5 NP_003465.3 2 530
NM_021641.4 NP_067673.2 2 530
GenBank mRNA AF023476.2 - 2 423
AF023477.2 - 2 423
AK127856.1 - 2 435
AY358878.1 - 2 427
BC060804.1 - 2 469 30348514
CB960379.1 - 2 495
DA863951.1 - 2 530
DR004947.1 - 2 426
KF444157.1 - 2 530

View Full Product Details