Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018404.2 | NP_060874.1 | 9 | 1144 | |
GenBank mRNA | AJ238994.1 | - | 9 | 1153 | |
AJ242782.1 | - | 9 | 1158 | ||
AJ272195.1 | - | 9 | 965 | ||
AK074196.1 | - | 8 | 1121 | ||
AK074204.1 | - | 9 | 972 | ||
AK222526.1 | - | 9 | 972 | ||
BC033758.1 | - | 9 | 957 | 5214358 | |
DQ288859.1 | - | 9 | 966 |