Gene Transcripts

Gene Symbol
SDK2
Entrez Gene ID
54549
Gene Name
sidekick cell adhesion molecule 2
Gene Aliases
Chromosome Location
Chr. 17: 73334384 - 73644482 on Build GRCh38
UniGene ID
Hs.435719
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144952.1 NP_001138424.1 21 2807
XM_011524914.2 XP_011523216.1 21 3191
XM_011524915.2 XP_011523217.1 21 3193
XM_011524916.2 XP_011523218.1 21 3201
GenBank mRNA AB040947.1 - 14 1838
BC045763.1 - 3 335
BC066363.1 - 13 1707

View Full Product Details