Gene Transcripts

Gene Symbol
ATG16L1
Entrez Gene ID
55054
Gene Name
autophagy related 16 like 1
Gene Aliases
APG16L, ATG16A, ATG16L, IBD10, WDR30
Chromosome Location
Chr. 2: 233251571 - 233295674 on Build GRCh38
UniGene ID
Hs.529322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190266.1 NP_001177195.1 15 1702
NM_001190267.1 NP_001177196.1 15 1700
NM_017974.3 NP_060444.3 14 1647
NM_030803.6 NP_110430.5 15 1704
NM_198890.2 NP_942593.2 11 1215
XM_005246082.2 XP_005246139.1 16 1692
XM_005246084.2 XP_005246141.1 13 1231
XM_005246086.2 XP_005246143.1 12 1180
XM_006712608.2 XP_006712671.1 15 1503
GenBank mRNA AK001690.1 - 14 1492
AK024453.1 - 13 2515
AK026330.1 - 2 182
AK027854.1 - 13 1229
AK074434.1 - 9 674
AK123876.1 - 12 1066
AK308400.1 - 14 1543
AY358182.1 - 14 1866
AY398617.1 - 15 1593
BC071846.1 - 11 1053
BC117337.1 - 15 1366
BC143900.1 - 14 1309
EF079889.1 - 14 1390
EF079890.1 - 11 958
EF560734.1 - 14 1517
JQ924062.1 - 14 1390

View Full Product Details