Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013313.4 | NP_037445.1 | 5 | 1764 | |
NR_130910.1 | 5 | 2148 | |||
GenBank mRNA | AB098734.1 | - | 5 | 1761 | |
BC034486.1 | - | 5 | 1623 | 4826997 | |
BQ422551.1 | - | 471 | |||
CR933630.1 | - | 4 | 5868 |