Gene Transcripts
- Gene Symbol
- CFAP61
- Entrez Gene ID
- 26074
- Gene Name
- cilia and flagella associated protein 61
- Gene Aliases
- C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
- Chromosome Location
- Chr. 20: 20052514 - 20360714 on Build GRCh38
- UniGene ID
- Hs.729168
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015585.3 |
NP_056400.3
|
24, 25 |
3329 |
|
XM_005260688.3 |
XP_005260745.1
|
24, 25 |
3307 |
|
XM_005260689.2 |
XP_005260746.1
|
24, 25 |
3292 |
|
XM_005260690.3 |
XP_005260747.1
|
23, 24 |
3232 |
|
XM_005260691.3 |
XP_005260748.1
|
23, 24 |
3229 |
|
XM_005260692.3 |
XP_005260749.1
|
23, 24 |
3214 |
|
XM_005260693.3 |
XP_005260750.1
|
24, 25 |
3307 |
|
XM_005260695.4 |
XP_005260752.1
|
19, 20 |
3618 |
|
XM_005260696.4 |
XP_005260753.1
|
15, 16 |
2191 |
|
XM_011529211.1 |
XP_011527513.1
|
23, 24 |
3165 |
|
XM_011529212.2 |
XP_011527514.1
|
24, 25 |
3307 |
|
XM_017027786.1 |
XP_016883275.1
|
24, 25 |
3247 |
|
XM_017027787.1 |
XP_016883276.1
|
24, 25 |
3232 |
|
XM_017027788.1 |
XP_016883277.1
|
23, 24 |
3214 |
|
XM_017027791.1 |
XP_016883280.1
|
14, 15 |
2089 |
|
XM_017027792.1 |
XP_016883281.1
|
15, 16 |
2131 |
|
XM_017027793.1 |
XP_016883282.1
|
14, 15 |
2098 |
|
XM_017027794.1 |
XP_016883283.1
|
14, 15 |
2162 |
|
GenBank mRNA |
AK057364.1 |
- |
7, 8 |
1014 |
|
AK304174.1 |
- |
25, 26 |
3190 |
|
BC028708.2 |
- |
24, 25 |
3291 |
4825941 |
View Full Product Details