Gene Transcripts
- Gene Symbol
- ANKFY1
- Entrez Gene ID
- 51479
- Gene Name
- ankyrin repeat and FYVE domain containing 1
- Gene Aliases
- ANKHZN, BTBD23, ZFYVE14
- Chromosome Location
- Chr. 17: 4163371 - 4264026 on Build GRCh38
- UniGene ID
- Hs.696087
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001257999.1 |
NP_001244928.1
|
2, 3 |
454 |
|
NM_016376.3 |
NP_057460.3
|
2, 3 |
249 |
|
XM_005256679.3 |
XP_005256736.1
|
2, 3 |
296 |
|
XM_011523926.1 |
XP_011522228.1
|
2, 3 |
352 |
|
XM_011523928.2 |
XP_011522230.1
|
2, 3 |
334 |
|
XM_017024733.1 |
XP_016880222.1
|
2, 3 |
549 |
|
GenBank mRNA |
AB033081.2 |
- |
2, 3 |
454 |
|
AB037360.1 |
- |
2, 3 |
217 |
|
AK025483.1 |
- |
2, 3 |
226 |
|
AK225170.1 |
- |
2, 3 |
226 |
|
AK292930.1 |
- |
2, 3 |
226 |
|
AK302712.1 |
- |
2, 3 |
235 |
|
BC052308.1 |
- |
2, 3 |
222 |
|
DA856905.1 |
- |
2, 3 |
249 |
|
View Full Product Details