Gene Transcripts

Gene Symbol
RTTN
Entrez Gene ID
25914
Gene Name
rotatin
Gene Aliases
MSSP
Chromosome Location
Chr. 18: 70003804 - 70205726 on Build GRCh38
UniGene ID
Hs.654809
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318520.1 NP_001305449.1 2, 3 282
NM_173630.3 NP_775901.3 2, 3 282
XM_006722434.3 XP_006722497.1 2, 3 282
XM_006722435.3 XP_006722498.1 2, 3 282
XM_011525902.2 XP_011524204.1 2, 3 282
XM_011525903.2 XP_011524205.1 2, 3 282
XM_011525904.2 XP_011524206.1 2, 3 282
XM_011525905.2 XP_011524207.1 2, 3 282
XM_011525907.2 XP_011524209.1 2, 3 282
XM_011525908.2 XP_011524210.1 2, 3 282
XM_017025693.1 XP_016881182.1 2, 3 282
XM_017025696.1 XP_016881185.1 2, 3 217
GenBank mRNA AK096404.1 - 2, 3 285
BC143723.1 - 2, 3 256
BF507420.1 - 1, 2 85

View Full Product Details