Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281443.1 | NP_001268372.1 | 3 | 167 | |
NM_007000.3 | NP_008931.1 | 3 | 167 | ||
XM_017026225.1 | XP_016881714.1 | 3 | 556 | ||
GenBank mRNA | AF085807.1 | - | 2 | 135 | |
BC107097.1 | - | 2 | 140 | 40015698 | |
BC107098.1 | - | 2 | 140 | 40015700 |