Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007224.3 | NP_009155.1 | 2 | 573 | |
GenBank mRNA | AK094231.1 | - | 3 | 778 | |
AK290949.1 | - | 2 | 511 | ||
AY358614.1 | - | 2 | 564 | ||
BC036679.1 | - | 2 | 567 | 5264190 | |
BC053581.1 | - | 2 | 497 | 5174266 |