Gene Transcripts

Gene Symbol
VAX1
Entrez Gene ID
11023
Gene Name
ventral anterior homeobox 1
Gene Aliases
MCOPS11
Chromosome Location
Chr. 10: 117128521 - 117138301 on Build GRCh38
UniGene ID
Hs.441536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199131.2 NP_954582.1 3, 4 707
GenBank mRNA AK127095.1 - 3, 4 704
BC101694.1 - 3, 4 491 8069200
BC101696.1 - 3, 4 491 8069202

View Full Product Details