Gene Transcripts
- Gene Symbol
- MED21
- Entrez Gene ID
- 9412
- Gene Name
- mediator complex subunit 21
- Gene Aliases
- SRB7, SURB7, hSrb7
- Chromosome Location
- Chr. 12: 27022522 - 27030673 on Build GRCh38
- UniGene ID
- Hs.286145
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004264.4 |
NP_004255.2
|
2, 3 |
213 |
|
NR_073464.1 |
|
2, 3 |
463 |
|
GenBank mRNA |
AK311838.1 |
- |
2, 3 |
157 |
|
BC008380.1 |
- |
2, 3 |
185 |
4041267 |
CR456999.1 |
- |
2, 3 |
155 |
|
DA420057.1 |
- |
2, 3 |
424 |
|
DB024509.1 |
- |
2, 3 |
213 |
|
KU178474.1 |
- |
2, 3 |
155 |
|
U46837.1 |
- |
2, 3 |
164 |
|
U52960.1 |
- |
2, 3 |
160 |
|
View Full Product Details