Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005736.3 | NP_005727.1 | 7 | 831 | |
GenBank mRNA | AK096085.1 | - | 1 | 643 | |
AK297362.1 | - | 7 | 668 | ||
AK297620.1 | - | 6 | 725 | ||
AK302072.1 | - | 6 | 720 | ||
AK312506.1 | - | 7 | 790 | ||
BC000693.2 | - | 7 | 767 | 3347881 | |
BC026016.2 | - | 7 | 733 | 3009822 | |
X82206.1 | - | 7 | 789 | ||
Z14978.1 | - | 7 | 771 |