Gene Transcripts

Gene Symbol
DHX16
Entrez Gene ID
8449
Gene Name
DEAH-box helicase 16
Gene Aliases
DBP2, DDX16, PRO2014, PRP8, PRPF2, Prp2
Chromosome Location
Chr. 6: 30653119 - 30673053 on Build GRCh38
UniGene ID
Hs.485060
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164239.1 NP_001157711.1 15 2221
NM_003587.4 NP_003578.2 15 2542
XM_011514938.2 XP_011513240.1 12 2335
XM_011514939.2 XP_011513241.1 12 1809
XM_011514940.2 XP_011513242.1 15 2750
XM_011514941.2 XP_011513243.1 7 834
GenBank mRNA AB001601.1 - 15 2469
AF119860.1 - 7 1014
AK302721.1 - 15 2215
AK312264.1 - 15 2481
BC008825.2 - 15 2493 4100320
BC009392.2 - 15 2493 4136069

View Full Product Details