Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318324.1 | NP_001305253.1 | 3 | 454 | |
NM_005085.3 | NP_005076.3 | 3 | 454 | ||
XM_005272216.3 | XP_005272273.1 | 3 | 454 | ||
XM_005272218.3 | XP_005272275.1 | 3 | 454 | ||
XM_005272219.3 | XP_005272276.1 | 3 | 454 | ||
XM_006717292.3 | XP_006717355.1 | 3 | 454 | ||
XM_011519053.2 | XP_011517355.1 | 3 | 454 | ||
XM_011519055.2 | XP_011517357.1 | 3 | 454 | ||
XM_017015167.1 | XP_016870656.1 | 3 | 454 | ||
GenBank mRNA | AB159230.1 | - | 3 | 313 | |
AK307573.1 | - | 3 | 426 | ||
BC012500.1 | - | 3 | 398 | ||
BC045620.1 | - | 3 | 398 | 5268515 | |
BC105998.1 | - | 3 | 429 | 30531438 | |
BM455135.1 | - | 421 | |||
D14689.1 | - | 3 | 365 | ||
DB062584.1 | - | 3 | 456 | ||
X64228.1 | - | 3 | 404 |