Gene Transcripts
- Gene Symbol
- TRIM54
- Entrez Gene ID
- 57159
- Gene Name
- tripartite motif containing 54
- Gene Aliases
- MURF, MURF-3, RNF30, muRF3
- Chromosome Location
- Chr. 2: 27282429 - 27307439 on Build GRCh38
- UniGene ID
- Hs.516036
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK300699.1 |
- |
|
1481 |
|
- Gene Symbol
- MLF2
- Entrez Gene ID
- 8079
- Gene Name
- myeloid leukemia factor 2
- Gene Aliases
- NTN4
- Chromosome Location
- Chr. 12: 6747992 - 6753470 on Build GRCh38
- UniGene ID
- Hs.524214
- Species
- Homo sapiens
View Full Product Details