Gene Transcripts

Gene Symbol
THBS3
Entrez Gene ID
7059
Gene Name
thrombospondin 3
Gene Aliases
TSP3
Chromosome Location
Chr. 1: 155195588 - 155209180 on Build GRCh38
UniGene ID
Hs.169875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252607.1 NP_001239536.1 14, 15 1900
NM_001252608.1 NP_001239537.1 13, 14 1567
NM_007112.4 NP_009043.1 15, 16 1927
NR_045553.1 14, 15 1911
XM_006711498.3 XP_006711561.1 15, 16 3486
GenBank mRNA AK074796.1 - 12, 13 1263
AK223278.1 - 15, 16 1841
AK298592.1 - 13, 14 1503
AK298803.1 - 11, 12 1235
AK298914.1 - 5, 6 577
AK314337.1 - 14, 15 1851
BC018786.1 - 6, 7 730
BC113846.1 - 14, 15 1829 40008346
BC113847.1 - 15, 16 1845 40008343
L38969.1 - 15, 16 1843

View Full Product Details