Gene Transcripts
- Gene Symbol
- PICALM
- Entrez Gene ID
- 8301
- Gene Name
- phosphatidylinositol binding clathrin assembly protein
- Gene Aliases
- CALM, CLTH, LAP
- Chromosome Location
- Chr. 11: 85957171 - 86069881 on Build GRCh38
- UniGene ID
- Hs.163893
- Species
- Homo sapiens
- Gene Symbol
- MLLT10
- Entrez Gene ID
- 8028
- Gene Name
- myeloid/lymphoid or mixed-lineage leukemia; translocated to, 10
- Gene Aliases
- AF10
- Chromosome Location
- Chr. 10: 21533349 - 21743630 on Build GRCh38
- UniGene ID
- Hs.30385
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF060929.1 |
- |
|
237 |
|
View Full Product Details