Gene Transcripts

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
Chromosome Location
Chr. 10: 109864766 - 109923553 on Build GRCh38
UniGene ID
Hs.390623
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324128.1 NP_001311057.1 13 1334
NM_001324131.1 NP_001311060.1 15 1531
NM_001324132.1 NP_001311061.1 14 1398
NM_001324133.1 NP_001311062.1 15 1487
NM_001324134.1 NP_001311063.1 13 1318
NM_001324135.1 NP_001311064.1 14 1436
NM_001324136.1 NP_001311065.1 15 1702
NM_020383.3 NP_065116.3 15 1487
XM_011540134.1 XP_011538436.1 15 1481
XM_017016613.1 XP_016872102.1 14 1337
XM_017016614.1 XP_016872103.1 16 1470
XM_017016615.1 XP_016872104.1 14 1354
GenBank mRNA AF195530.1 - 14 1314
AF272981.1 - 13 1238
AK093693.1 - 13 1292
AK095447.1 - 1 1707
AK223513.1 - 14 1298
AK289436.1 - 15 1390
BC005126.2 - 14 1276 3836436
BC007579.1 - 15 1378 3139868
BC013417.2 - 11 988
BX537682.1 - 11 3332
CR456922.1 - 13 1238
X95762.1 - 13 1238

View Full Product Details