Gene Transcripts

Gene Symbol
XPNPEP1
Entrez Gene ID
7511
Gene Name
X-prolyl aminopeptidase 1
Gene Aliases
APP1, SAMP, XPNPEP, XPNPEPL, XPNPEPL1
Chromosome Location
Chr. 10: 109864766 - 109923553 on Build GRCh38
UniGene ID
Hs.390623
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167604.1 NP_001161076.1 16, 17 1573
NM_001324128.1 NP_001311057.1 15, 16 1492
NM_001324131.1 NP_001311060.1 17, 18 1689
NM_001324132.1 NP_001311061.1 16, 17 1556
NM_001324133.1 NP_001311062.1 17, 18 1645
NM_001324134.1 NP_001311063.1 15, 16 1476
NM_001324135.1 NP_001311064.1 16, 17 1594
NM_001324136.1 NP_001311065.1 17, 18 1860
NM_020383.3 NP_065116.3 17, 18 1645
XM_017016613.1 XP_016872102.1 16, 17 1495
XM_017016614.1 XP_016872103.1 18, 19 1628
XM_017016615.1 XP_016872104.1 16, 17 1512
GenBank mRNA AF195530.1 - 16, 17 1472
AF272981.1 - 15, 16 1396
AK093693.1 - 15, 16 1450
AK223513.1 - 16, 17 1456
AK289436.1 - 17, 18 1548
AL833411.1 - 16, 17 1479
BC005126.2 - 16, 17 1434 3836436
BC007579.1 - 17, 18 1536 3139868
BC013417.2 - 13, 14 1146
BX537682.1 - 13, 14 3490
CR456922.1 - 15, 16 1396
X95762.1 - 15, 16 1396

View Full Product Details