Gene Transcripts

Gene Symbol
ARSG
Entrez Gene ID
22901
Gene Name
arylsulfatase G
Gene Aliases
Chromosome Location
Chr. 17: 68259089 - 68452000 on Build GRCh38
UniGene ID
Hs.437249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267727.1 NP_001254656.1 5, 6 1192
NM_014960.4 NP_055775.2 5, 6 1363
XM_005257170.3 XP_005257227.1 5, 6 1376
XM_005257172.3 XP_005257229.1 5, 6 1425
XM_006721777.3 XP_006721840.2 5, 6 1390
XM_006721779.3 XP_006721842.1 5, 6 1417
XM_011524535.2 XP_011522837.1 5, 6 1388
XM_011524536.2 XP_011522838.1 5, 6 1218
XM_011524537.1 XP_011522839.1 5, 6 1139
XM_011524538.2 XP_011522840.1 5, 6 1390
XM_011524540.2 XP_011522842.1 5, 6 1413
XM_011524541.2 XP_011522843.1 5, 6 1456
XM_011524542.2 XP_011522844.1 5, 6 1456
XM_011524543.2 XP_011522845.1 5, 6 1456
XM_011524544.2 XP_011522846.1 5, 6 1456
XM_011524545.2 XP_011522847.1 5, 6 1414
XM_017024360.1 XP_016879849.1 5, 6 1228
XM_017024361.1 XP_016879850.1 5, 6 1376
XM_017024362.1 XP_016879851.1 5, 6 1315
XM_017024363.1 XP_016879852.1 5, 6 1221
XM_017024364.1 XP_016879853.1 5, 6 1217
XM_017024365.1 XP_016879854.1 5, 6 1205
XM_017024366.1 XP_016879855.1 5, 6 1416
XM_017024367.1 XP_016879856.1 5, 6 1218
XM_017024368.1 XP_016879857.1 5, 6 1218
GenBank mRNA AB023218.1 - 4, 5 1025
AY358380.1 - 5, 6 1363
BC012375.1 - 4, 5 606 3882163

View Full Product Details