Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003958.3 | NP_003949.1 | 3 | 662 | |
NM_183078.2 | NP_898901.1 | 3 | 662 | ||
NR_046399.1 | 3 | 768 | |||
XM_006715241.2 | XP_006715304.1 | 3 | 662 | ||
XM_006715242.2 | XP_006715305.1 | 3 | 662 | ||
XM_017011462.1 | XP_016866951.1 | 3 | 513 | ||
XM_017011463.1 | XP_016866952.1 | 3 | 518 | ||
XM_017011464.1 | XP_016866953.1 | 3 | 667 | ||
GenBank mRNA | AB014546.1 | - | 3 | 581 | |
AF334675.1 | - | 3 | 469 | ||
AK022075.1 | - | 3 | 563 | ||
AK222765.1 | - | 3 | 580 | ||
AK298319.1 | - | 3 | 686 | ||
AK308840.1 | - | 3 | 569 | ||
AL711279.1 | - | 3 | 620 | ||
BC007517.2 | - | 3 | 591 | 3028746 | |
BT007446.1 | - | 3 | 469 |