Gene Transcripts

Gene Symbol
ALPK3
Entrez Gene ID
57538
Gene Name
alpha kinase 3
Gene Aliases
MAK, MIDORI
Chromosome Location
Chr. 15: 84816631 - 84873482 on Build GRCh38
UniGene ID
Hs.459183
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020778.4 NP_065829.3 1, 2 921
XM_017022445.1 XP_016877934.1 1, 2 970
GenBank mRNA AY044449.1 - 1, 2 921

View Full Product Details