Gene Transcripts

Gene Symbol
ATG101
Entrez Gene ID
60673
Gene Name
autophagy related 101
Gene Aliases
C12orf44
Chromosome Location
Chr. 12: 52069974 - 52077495 on Build GRCh38
UniGene ID
Hs.9911
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098673.1 NP_001092143.1 4 840
NM_021934.4 NP_068753.2 4 1011
GenBank mRNA AF218031.1 - 4 951
AK021835.1 - 4 1009
BC005151.2 - 4 963 3508173
BC009937.2 - 4 945 4303452

View Full Product Details