Gene Transcripts

Gene Symbol
CEP95
Entrez Gene ID
90799
Gene Name
centrosomal protein 95
Gene Aliases
CCDC45
Chromosome Location
Chr. 17: 64506736 - 64537951 on Build GRCh38
UniGene ID
Hs.569713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316990.1 NP_001303919.1 3 569
NM_138363.2 NP_612372.1 3 569
NR_133644.1 3 569
XM_005257779.3 XP_005257836.1 3 545
XM_005257780.2 XP_005257837.1 3 294
XM_006722166.3 XP_006722229.1 3 311
XM_011525458.2 XP_011523760.1 3 545
XM_017025287.1 XP_016880776.1 3 313
XM_017025288.1 XP_016880777.1 3 311
GenBank mRNA AK057326.1 - 3 543
AK290587.1 - 3 309
AK297409.1 - 3 270
AK316257.1 - 3 313
AL832822.1 - 3 220
BC009518.1 - 3 265 3633884
BP357015.1 - 3 569

View Full Product Details