Gene Transcripts

Gene Symbol
C15orf32
Entrez Gene ID
145858
Gene Name
chromosome 15 open reading frame 32
Gene Aliases
Chromosome Location
Chr. 15: 92465762 - 92501117 on Build GRCh38
UniGene ID
Hs.367879
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301106.1 NP_001288035.1 1 833
NM_153040.2 NP_694585.1 1 833
XM_011521257.2 XP_011519559.1 1 6748
GenBank mRNA AK057393.1 - 1 834
BC109252.2 - 1 589 40023840
BC109253.1 - 1 587 40023844

View Full Product Details