Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136026.2 | NP_001129498.1 | 10 | 1282 | |
NM_001136232.2 | NP_001129704.1 | 9 | 1136 | ||
NM_030673.3 | NP_109598.2 | 10 | 1138 | ||
NM_183352.2 | NP_899195.1 | 9 | 1104 | ||
XM_005265379.2 | XP_005265436.1 | 10 | 1155 | ||
XM_017007019.1 | XP_016862508.1 | 10 | 1192 | ||
GenBank mRNA | AB209554.1 | - | 9 | 3278 | |
AF052155.1 | - | 9 | 1136 | ||
AK095629.1 | - | 7 | 2963 | ||
AK223019.1 | - | 9 | 989 | ||
AK301999.1 | - | 10 | 1206 | ||
BC002634.2 | - | 10 | 1019 | 3605545 | |
BC006167.1 | - | 6 | 796 | ||
BC091506.1 | - | 9 | 1014 | 30408210 | |
DB564012.1 | - | 1 | 80 | ||
L09260.1 | - | 9 | 981 |