Gene Transcripts
- Gene Symbol
- SLC28A2
- Entrez Gene ID
- 9153
- Gene Name
- solute carrier family 28 member 2
- Gene Aliases
- CNT2, HCNT2, HsT17153, SPNT1
- Chromosome Location
- Chr. 15: 45252230 - 45275934 on Build GRCh38
- UniGene ID
- Hs.367833
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004212.3 |
NP_004203.2
|
5, 6 |
504 |
|
XM_011522198.2 |
XP_011520500.1
|
5, 6 |
524 |
|
XM_011522199.1 |
XP_011520501.1
|
5, 6 |
475 |
|
XM_011522200.2 |
XP_011520502.1
|
5, 6 |
475 |
|
XM_011522201.2 |
XP_011520503.1
|
5, 6 |
475 |
|
GenBank mRNA |
AF036109.1 |
- |
4, 5 |
453 |
|
AK222709.1 |
- |
5, 6 |
504 |
|
AK291974.1 |
- |
5, 6 |
475 |
|
BC093737.1 |
- |
4, 5 |
467 |
7939582 |
BC112047.1 |
- |
4, 5 |
479 |
8327515 |
U84392.1 |
- |
5, 6 |
498 |
|
View Full Product Details