Gene Transcripts

Gene Symbol
TM4SF1
Entrez Gene ID
4071
Gene Name
transmembrane 4 L six family member 1
Gene Aliases
H-L6, L6, M3S1, TAAL6
Chromosome Location
Chr. 3: 149369018 - 149377860 on Build GRCh38
UniGene ID
Hs.351316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014220.2 NP_055035.1 5 1640
XM_017006385.1 XP_016861874.1 5 1660
GenBank mRNA AK093829.1 - 2 3068
AK130073.1 - 5 1552
AK130271.1 - 5 1634
AL832780.1 - 1 7114
BC008442.1 - 5 1508 4101110
BC034145.1 - 5 1600 4618019
BI494959.1 - 1 520
X75684.1 - 1 100

View Full Product Details