Gene Transcripts

Gene Symbol
IL17A
Entrez Gene ID
3605
Gene Name
interleukin 17A
Gene Aliases
CTLA-8, CTLA8, IL-17, IL-17A, IL17
Chromosome Location
Chr. 6: 52186387 - 52190638 on Build GRCh38
UniGene ID
Hs.41724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002190.2 NP_002181.1 3 983
GenBank mRNA U32659.1 - 3 991
Z58820.1 - 3 964

View Full Product Details