Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004273.4 | NP_004264.2 | 3 | 4116 | |
XM_006718075.3 | XP_006718138.1 | 3 | 4349 | ||
XM_011540369.2 | XP_011538671.1 | 3 | 6042 | ||
GenBank mRNA | AB017915.1 | - | 3 | 4114 | |
BG325090.1 | - | 1 | 298 |