Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001794.4 | NP_001785.2 | 2 | 218 | |
GenBank mRNA | AK315192.1 | - | 2 | 172 | |
BC101651.1 | - | 2 | 218 | 8069157 | |
BC112150.1 | - | 2 | 218 | 8327618 | |
L34059.1 | - | 2 | 144 |