Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008844.2 | NP_001008844.1 | 1 | 465 | |
NM_001319034.1 | NP_001305963.1 | 1 | 465 | ||
NM_004415.3 | NP_004406.2 | 1 | 465 | ||
GenBank mRNA | AB209992.1 | - | 1 | 423 | |
AB621818.1 | - | 1 | 370 | ||
BC140802.1 | - | 1 | 398 | 9021670 | |
HM151899.1 | - | 1 | 403 | ||
M77830.3 | - | 1 | 403 |