Gene Transcripts
- Gene Symbol
- CASP10
- Entrez Gene ID
- 843
- Gene Name
- caspase 10
- Gene Aliases
- ALPS2, FLICE2, MCH4
- Chromosome Location
- Chr. 2: 201182881 - 201229406 on Build GRCh38
- UniGene ID
- Hs.5353
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001206524.1 |
NP_001193453.1
|
4, 5 |
1003 |
|
NM_001206542.1 |
NP_001193471.1
|
4, 5 |
1003 |
|
NM_001230.4 |
NP_001221.2
|
4, 5 |
1003 |
|
NM_001306083.1 |
NP_001293012.1
|
4, 5 |
1020 |
|
NM_032974.4 |
NP_116756.2
|
4, 5 |
1003 |
|
NM_032976.3 |
NP_116758.1
|
4, 5 |
1003 |
|
NM_032977.3 |
NP_116759.2
|
4, 5 |
1003 |
|
XM_005246907.2 |
XP_005246964.1
|
4, 5 |
789 |
|
XM_006712796.3 |
XP_006712859.1
|
4, 5 |
1016 |
|
XM_011511990.2 |
XP_011510292.1
|
4, 5 |
789 |
|
GenBank mRNA |
AF111344.1 |
- |
3, 4 |
585 |
|
AF111345.1 |
- |
3, 4 |
585 |
|
AJ487678.1 |
- |
4, 5 |
709 |
|
AJ487679.1 |
- |
4, 5 |
706 |
|
AK225475.1 |
- |
4, 5 |
741 |
|
AK304859.1 |
- |
3, 4 |
666 |
|
AK310746.1 |
- |
4, 5 |
789 |
|
AY690601.3 |
- |
4, 5 |
741 |
|
BC042844.1 |
- |
4, 5 |
739 |
5176698 |
KU177959.1 |
- |
3, 4 |
585 |
|
KU177960.1 |
- |
3, 4 |
585 |
|
U60519.1 |
- |
4, 5 |
732 |
|
U86214.1 |
- |
4, 5 |
779 |
|
View Full Product Details