Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000662.7 | NP_000653.3 | 3 | 1082 | |
NM_001160170.3 | NP_001153642.1 | 5 | 1616 | ||
NM_001160171.3 | NP_001153643.1 | 4 | 1466 | ||
NM_001160172.3 | NP_001153644.1 | 5 | 1387 | ||
NM_001160173.3 | NP_001153645.1 | 4 | 1232 | ||
NM_001160174.2 | NP_001153646.1 | 1 | 1299 | ||
NM_001160175.3 | NP_001153647.1 | 5 | 1350 | ||
NM_001160176.3 | NP_001153648.1 | 4 | 1200 | ||
NM_001160179.2 | NP_001153651.1 | 5 | 1286 | ||
NM_001291962.1 | NP_001278891.1 | 6 | 1404 | ||
XM_006716410.3 | XP_006716473.1 | 3 | 1659 | ||
XM_011544687.1 | XP_011542989.1 | 6 | 1745 | ||
XM_011544688.1 | XP_011542990.1 | 5 | 1595 | ||
XM_011544689.2 | XP_011542991.1 | 2 | 1444 | ||
XM_017013947.1 | XP_016869436.1 | 8 | 1981 | ||
GenBank mRNA | AJ581142.1 | - | 1 | 176 | |
AJ581143.1 | - | 1 | 176 | ||
AK290912.1 | - | 3 | 1557 | ||
BC013732.1 | - | 4 | 1439 | ||
BC047666.1 | - | 3 | 1061 | 6059458 | |
BX647521.1 | - | 4 | 1175 | ||
D90041.1 | - | 1 | 1065 | ||
S78829.1 | - | 1 | 60 |