Gene Transcripts

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
Chromosome Location
Chr. 2: 110637698 - 110678107 on Build GRCh38
UniGene ID
Hs.469649
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278616.1 NP_001265545.1 6 658
NM_001278617.1 NP_001265546.1 7 718
NM_004336.4 NP_004327.1 7 718
GenBank mRNA AF043294.2 - 7 638
AF046078.1 - 7 655
AF047471.1 - 7 641
AF053305.1 - 7 669
AK023540.1 - 7 674
AK225317.1 - 7 674
AK291221.1 - 7 671
AK302418.1 - 6 614
BC017038.1 - 7 639
BC028201.1 - 7 639 5205385

View Full Product Details