Gene Transcripts
- Gene Symbol
- CATSPERG
- Entrez Gene ID
- 57828
- Gene Name
- cation channel sperm associated auxiliary subunit gamma
- Gene Aliases
- C19orf15
- Chromosome Location
- Chr. 19: 38335563 - 38370949 on Build GRCh38
- UniGene ID
- Hs.324335
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021185.4 |
NP_067008.3
|
10, 11 |
1280 |
|
XM_005259114.4 |
XP_005259171.1
|
10, 11 |
1376 |
|
XM_005259115.4 |
XP_005259172.1
|
10, 11 |
1299 |
|
XM_006723308.3 |
XP_006723371.1
|
10, 11 |
1896 |
|
XM_006723310.2 |
XP_006723373.1
|
10, 11 |
1280 |
|
XM_006723311.2 |
XP_006723374.1
|
10, 11 |
1301 |
|
XM_011527174.2 |
XP_011525476.1
|
10, 11 |
1290 |
|
XM_011527175.2 |
XP_011525477.1
|
10, 11 |
1196 |
|
XM_011527176.2 |
XP_011525478.1
|
10, 11 |
1280 |
|
XM_011527177.2 |
XP_011525479.1
|
10, 11 |
1160 |
|
XM_011527178.2 |
XP_011525480.1
|
11, 12 |
1642 |
|
XM_011527179.1 |
XP_011525481.1
|
10, 11 |
1280 |
|
XM_011527182.2 |
XP_011525484.1
|
9, 10 |
1024 |
|
XM_017027054.1 |
XP_016882543.1
|
8, 9 |
893 |
|
GenBank mRNA |
AK128220.1 |
- |
10, 11 |
1063 |
|
AK310460.1 |
- |
6, 7 |
610 |
|
AL133023.1 |
- |
10, 11 |
1896 |
|
BC094785.1 |
- |
10, 11 |
1200 |
|
BC109035.1 |
- |
6, 7 |
679 |
40028284 |
View Full Product Details